1,2-Cyclohexanedione ≥97.0%
Supplier: TCI America
C0728-5G
C0728-25G
TCC0728-005GEA
117.33
CAD
TCC0728-005G
TCC0728-025G
1,2-Cyclohexanedione ≥97.0%
1,2-Cyclohexanedione
CAS Number: 765-87-7
MDL Number: MFCD00001648
Molecular Formula: C6H8O2
Molecular Weight: 112.13
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 194
Flash Point (°C): 84
Freezing point (°C): 38
Formula:
C₆H₈O₂ MW: 112.13 g/mol Boiling Pt: 194 °C Melting Pt: 33…35 °C Density: 1.1305 g/cm³ (20 °C) |
MDL Number:
MFCD00001648 CAS Number: 765-87-7 EINECS: 212-155-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...