(R)-(-)-Carvone ≥99.0%

Supplier: TCI America

C0703-25ML C0703-500ML C0703-100ML
TCC0703-025MLEA 72.13 CAD
TCC0703-025ML TCC0703-500ML TCC0703-100ML
(R)-(-)-Carvone ≥99.0%
(R)-(-)-Carvone

CAS Number: 6485-40-1
MDL Number: MFCD00001578
Molecular Formula: C10H14O
Molecular Weight: 150.22
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 231
Flash Point (°C): 88
Specific Gravity (20/20): 0.96
Specific rotation [a]20/D: -61.5 deg (neat)


Product Categories: Bioscience, Monocyclic Monoterpenes
Other Product Names / Synonyms: (R)-(-)-p-Mentha-6,8-dien-2-one

Formula: C₁₀H₁₄O
Boiling Pt: 231 °C
Melting Pt: 25.2 °C
Density: 0.96 (20 °C)
Flash Pt: 88 °C
MDL Number: MFCD00001578
CAS Number: 6485-40-1
EINECS: 229-352-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=556b0c1e994cdf39b50b3a778e160b1dfb02c08a&vid=556b0c1e994cdf39b50b3a778e160b1dfb02c08a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR