ε-Hexalactone ≥99.0%
Supplier: TCI America
Synonyms:
ε-Caprolactone, Oxepan-2-one, Hexan-6-olide
CAS Number: 502-44-3
MDL Number: MFCD00003267
Molecular Formula: C6H10O2
Molecular Weight: 114.14
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 140
Melting point (°C): -1
Flash Point (°C): 115
Specific Gravity (20/20): 1.08
Product Categories: Materials Chemistry, Monomers, Bioscience, Lipids, Monomers for Synthetic Polymers (for DDS)
Other Product Names / Synonyms: epsilon-Hexalactone, epsilon-Hexanolactone
Formula:
C₆H₁₀O₂ MW: 114.14 g/mol Boiling Pt: 140 °C Melting Pt: –1 °C Density: 1.08 (20 °C) Flash Pt: 115 °C |
MDL Number:
MFCD00003267 CAS Number: 502-44-3 EINECS: 207-938-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...