N-(Ethoxycarbonyl)phthalimide ≥98.0%

Supplier: TCI America

Synonyms: Ethyl 1,3-dioxoisoindoline 2-carboxylate, N-Carbethoxyphthalimide

C0683-500G C0683-25G C0683-100G
TCC0683-500GEA 512.39 CAD
TCC0683-500G TCC0683-025G TCC0683-100G
N-(Ethoxycarbonyl)phthalimide ≥98.0%
N-(Ethoxycarbonyl)phthalimide

[for Peptide Synthesis]
CAS Number: 22509-74-6
MDL Number: MFCD00005893
Molecular Formula: C11H9NO4
Molecular Weight: 219.20
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 87


Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis), Peptide Synthesis, Bioscience, Polyamine Synthesis Reagents
Other Product Names / Synonyms: N-Carbethoxyphthalimide

Formula: C₁₁H₉NO₄
MW: 219.20 g/mol
Melting Pt: 87 °C
MDL Number: MFCD00005893
CAS Number: 22509-74-6
EINECS: 245-048-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f81d5403a0bd74dd1896e07ef05db7a94db9161&vid=431611601aec115a19ebb9465f4f16e937f3b894&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR