1,3-Cyclohexanedione ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Cyclohexane-1,3-dione

C0682-25G C0682-250G
TCC0682-025GEA 43.29 CAD
TCC0682-025G TCC0682-250G
1,3-Cyclohexanedione ≥98.0% (by GC, titration analysis)
1,3-Cyclohexanedione

CAS Number: 504-02-9
MDL Number: MFCD00001585
Molecular Formula: C6H8O2
Molecular Weight: 112.13
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 105
Storage Temperature: 0-10°C

Formula: C₆H₈O₂
MW: 112.13 g/mol
Boiling Pt: 235 °C (1013 hPa)
Melting Pt: 105 °C
Density: 1.25 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00001585
CAS Number: 504-02-9
EINECS: 207-980-0
Merck Index: 13,03200
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7bef83303d232cf8943574e6c014247e0540a9f&vid=7dc23c922f28bbb9f1417597662c8ea45d14118d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR