Cloxiquina ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
5-Chloro-8-quinolinol, 5-Chloroquinolin-8-ol, 5-Chloro-8-hydroxyquinoline
C0645-25G
C0645-250G
TCC0645-025GEA
58.46
CAD
TCC0645-025G
TCC0645-250G
Cloxiquina ≥98.0% (by GC, titration analysis)
Cloxiquina
CAS Number: 130-16-5
MDL Number: MFCD00006788
Molecular Formula: C9H6ClNO
Molecular Weight: 179.60
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 127
Product Categories: Materials Chemistry, Ligands for Functional Metal Complexes
Other Product Names / Synonyms: 5-Chlorooxine, 5-Chloro-8-quinolinol, 5-Chloroquinophenol
Formula:
C₉H₆ClNO MW: 179.61 g/mol Boiling Pt: 349 °C (1013 hPa) Melting Pt: 127 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006788 CAS Number: 130-16-5 EINECS: 204-978-1 Merck Index: 12,02483 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...