4-Chloro-3,5-dinitrobenzoic acid ≥98.0%

Supplier: TCI America

Synonyms: 3,5-Dinitro-4-chlorobenzoic acid

C0607-25G C0607-250G
TCC0607-025GEA 122.29 CAD
TCC0607-025G TCC0607-250G
4-Chloro-3,5-dinitrobenzoic acid ≥98.0%
4-Chloro-3,5-dinitrobenzoic acid

CAS Number: 118-97-8
MDL Number: MFCD00007080
Molecular Formula: C7H3ClN2O6
Molecular Weight: 246.56
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 160
Flash Point (°C): 186

Formula: C₇H₃ClN₂O₆
Boiling Pt: 240 °C (12 mmHg)
Melting Pt: 160 °C
Flash Pt: 186 °C
Storage Temperature: Ambient
MDL Number: MFCD00007080
CAS Number: 118-97-8
EINECS: 204-290-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1dc9209195d4484884850976d5664d767658b30f&vid=2ac7b751d3ba24634e5398b877057ef18d0c7217&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR