Benzyl carbamate ≥97.0% (by total nitrogen basis)
Supplier: TCI America
C0590-25G
C0590-250G
TCC0590-025GEA
85.32
CAD
TCC0590-025G
TCC0590-250G
Benzyl carbamate ≥97.0% (by total nitrogen basis)
Benzyl carbamate
CAS Number: 621-84-1
MDL Number: MFCD00007965
Molecular Formula: C8H9NO2
Molecular Weight: 151.17
Purity/Analysis Method: >97.0% (N)
Form: Crystal
Melting point (°C): 87
Other Product Names / Synonyms: Carbamic Acid Benzyl Ester
Formula:
C₈H₉NO₂ Melting Pt: 87 °C |
MDL Number:
MFCD00007965 CAS Number: 621-84-1 EINECS: 210-710-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...