N-Cbz-glycine(Z-Gly-OH) ≥98.0%

Supplier: TCI America

Synonyms: N-Carbobenzyloxyglycine, N-Benzyloxycarbonylglycine, Z-Gly-OH, 2-(((Benzyloxy)carbonyl)amino)acetic acid

C0575-25G C0575-500G
TCC0575-025GEA 44.35 CAD
TCC0575-025G TCC0575-500G
N-Cbz-glycine(Z-Gly-OH) ≥98.0%
N-Cbz-Glycine (Z-Gly-OH)

CAS Number: 1138-80-3
MDL Number: MFCD00002691
Molecular Formula: C10H11NO4
Molecular Weight: 209.20
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 120


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Cbz-glycine, Z-Gly-OH

Formula: C₁₀H₁₁NO₄
MW: 209.2 g/mol
Melting Pt: 120 °C
MDL Number: MFCD00002691
CAS Number: 1138-80-3
EINECS: 214-516-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f1e9f2efce28287bf1af333f6513668bafb354dd&vid=f1e9f2efce28287bf1af333f6513668bafb354dd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR