4-Chlorobenzamide ≥98.0%

Supplier: TCI America

C0563-25G C0563-250G
TCC0563-025GEA 138.88 CAD
TCC0563-025G TCC0563-250G
4-Chlorobenzamide ≥98.0%
4-Chlorobenzamide

CAS Number: 619-56-7
MDL Number: MFCD00007993
Molecular Formula: C7H6ClNO
Molecular Weight: 155.58
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 179

Formula: C₇H₆ClNO
Boiling Pt: 314 °C (1013 hPa)
Melting Pt: 179 °C
MDL Number: MFCD00007993
CAS Number: 619-56-7
EINECS: 210-601-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=847b52a36a23e39b2233efd3be213750731e4efb&vid=847b52a36a23e39b2233efd3be213750731e4efb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR