3-Chlorobenzamide 98,0
Supplier: TCI America
C0562-250G
C0562-25G
TCC0562-250GEA
609.88
CAD
TCC0562-250G
TCC0562-025G
3-Chlorobenzamide 98,0
3-Chlorobenzamide
CAS Number: 618-48-4
MDL Number: MFCD00014799
Molecular Formula: C7H6ClNO
Molecular Weight: 155.58
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 134
Formula:
C₇H₆ClNO MW: 155.58 g/mol Melting Pt: 134 °C |
MDL Number:
MFCD00014799 CAS Number: 618-48-4 EINECS: 210-554-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...