Cyclohexanone oxime ≥98.0%

Supplier: TCI America
Warning

C0490-500G C0490-25G
TCC0490-500GEA 518.24 CAD
TCC0490-500G TCC0490-025G
Cyclohexanone oxime ≥98.0%
Cyclohexanone oxime

CAS Number: 100-64-1
MDL Number: MFCD00001660
Molecular Formula: C6H11NO
Molecular Weight: 113.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 210
Melting point (°C): 90
Flash Point (°C): 103

Formula: C₆H₁₁NO
MW: 113.16 g/mol
Boiling Pt: 206
Melting Pt: 90 °C
Density: 1.13 g/cm³ (20 °C)
Flash Pt: 103 °C
MDL Number: MFCD00001660
CAS Number: 100-64-1
EINECS: 202-874-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=97c40d9f54f3ee927f01c1c021cc863e9d8429af&vid=97c40d9f54f3ee927f01c1c021cc863e9d8429af&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR