Sodium cyclamate ≥98.0% (by total nitrogen basis)

Supplier: TCI America
Warning

C0487-25G C0487-500G
TCC0487-025GEA 33.18 CAD
TCC0487-025G TCC0487-500G
Sodium cyclamate ≥98.0% (by total nitrogen basis)
Sodium cyclamate

CAS Number: 139-05-9
MDL Number: MFCD00003827
Molecular Formula: C6H13NO3S
Molecular Weight: 201.22
Purity/Analysis Method: >98.0% (N)
Form: Crystal


Product Categories: Bioscience, Obesity/Diabete Research Reagents
Other Product Names / Synonyms: N-Cyclohexanesulfamic Acid Sodium Salt, N-Cyclohexylsulfamic Acid Sodium Salt, Sodium N-Cyclohexanesulfamate, Sodium Cyclamate, Cyclamic Acid Sodium Salt

Formula: C₆H₁₂NNaO₃S
Melting Pt: 265 °C
MDL Number: MFCD00003827
CAS Number: 139-05-9
EINECS: 205-348-9
Merck Index: 12,02770
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b1d84ec481c98cca7c691feb76c32931e4f02907&vid=b1d84ec481c98cca7c691feb76c32931e4f02907&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR