1,2-Cyclohexanedione dioxime ≥98.0% (by gravimetric analysis)

Supplier: TCI America

C0483-25G C0483-1G C0483-5G
TCC0483-025GEA 910.08 CAD
TCC0483-025G TCC0483-001G TCC0483-5G
1,2-Cyclohexanedione dioxime ≥98.0% (by gravimetric analysis)
1,2-Cyclohexanedione dioxime

CAS Number: 492-99-9
MDL Number: MFCD00001663
Molecular Formula: C6H10N2O2
Molecular Weight: 142.16
Purity/Analysis Method: >98.0% (W)
Form: Crystal
Melting point (°C): 188


Other Product Names / Synonyms: 1,2-Cyclohexanedione Dioxime

Formula: C₆H₁₀N₂O₂
MW: 142.16 g/mol
Melting Pt: 188 °C
MDL Number: MFCD00001663
CAS Number: 492-99-9
EINECS: 207-769-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae3099b498ce1c5cc52497c0142d92d63523deab&vid=ae3099b498ce1c5cc52497c0142d92d63523deab&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (W)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR