1,4-Cyclohexanediol ≥99.0%

Supplier: TCI America

Synonyms: Cyclohexane-1,4-diol

C0482-100G C0482-25G C0482-500G
TCC0482-100GEA 85.32 CAD
TCC0482-100G TCC0482-25G TCC0482-500G
1,4-Cyclohexanediol ≥99.0%
1,4-Cyclohexanediol

CAS Number: 556-48-9
MDL Number: MFCD00001448
Molecular Formula: C6H12O2
Molecular Weight: 116.16
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 132
Melting point (°C): 101
Flash Point (°C): 65


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,4-Dihydroxycyclohexane, Quinitol

Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 132 °C
Melting Pt: 101 °C
Density: 1.17 g/cm³ (21.85 °C)
Flash Pt: 65 °C
MDL Number: MFCD00001448
CAS Number: 556-48-9
EINECS: 209-126-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9bccbf43aec6358dbb03aedfcc2dacb53d009851&vid=92d1d1be2985c5eefe82c01a6c8bc1f784258ebb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR