1,3-Cyclohexanediol ≥98.0%
Supplier: TCI America
Synonyms:
Cyclohexane-1,3-diol
C0481-25G
C0481-500G
TCC0481-025GEA
71.1
CAD
TCC0481-025G
TCC0481-500G
1,3-Cyclohexanediol ≥98.0%
1,3-Cyclohexanediol
CAS Number: 504-01-8
MDL Number: MFCD00039458
Molecular Formula: C6H12O2
Molecular Weight: 116.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 247
Flash Point (°C): 113
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,3-Dihydroxycyclohexane
Formula:
C₆H₁₂O₂ MW: 116.16 g/mol Boiling Pt: 247 °C Melting Pt: 102 °C Flash Pt: 113 °C |
MDL Number:
MFCD00039458 CAS Number: 504-01-8 EINECS: 207-979-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...