1,2-Cyclohexanediol ≥98.0%
Supplier: TCI America
Synonyms:
Cyclohexane-1,2-diol
C0480-500G
C0480-25G
C0480-100G
TCC0480-500GEA
1162.88
CAD
TCC0480-500G
TCC0480-025G
TCC0480-100G
1,2-Cyclohexanediol ≥98.0%
1,2-Cyclohexanediol
CAS Number: 931-17-9
MDL Number: MFCD00003861
Molecular Formula: C6H12O2
Molecular Weight: 116.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 236
Melting point (°C): 104
Flash Point (°C): 110
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,2-Dihydroxycyclohexane
Formula:
C₆H₁₂O₂ MW: 116.16 g/mol Boiling Pt: 236 °C Melting Pt: 104 °C Density: 0.948 g/cm³ (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00003861 CAS Number: 931-17-9 EINECS: 213-229-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...