1,4-Bis(hydroxymethyl)cyclohexane (mixture of cis and trans isomers) ≥99.0%
Supplier: TCI America
C0479-25G
C0479-500G
TCC0479-025GEA
39.5
CAD
TCC0479-025G
TCC0479-500G
1,4-Bis(hydroxymethyl)cyclohexane (mixture of cis and trans isomers) ≥99.0%
1,4-Bis(hydroxymethyl)cyclohexane (mixture of cis and trans isomers)
CAS Number: 105-08-8
MDL Number: MFCD00001512
Molecular Formula: C8H16O2
Molecular Weight: 144.21
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 162
Flash Point (°C): 166
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,4-Bis(hydroxymethyl)cyclohexane, alpha,alpha'-Dihydroxy-1,4-dimethylcyclohexane
Formula:
C₈H₁₆O₂ Boiling Pt: 162 °C Flash Pt: 166 °C |
MDL Number:
MFCD00001512 CAS Number: 105-08-8 EINECS: 203-268-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...