trans-1,2-Cyclohexanedicarboxylic acid ≥98.0%

Supplier: TCI America

Synonyms: trans-Cyclohexane-1,2-dicarboxylic acid

C0474-500G C0474-25G
TCC0474-500GEA 1924.44 CAD
TCC0474-500G TCC0474-025G
trans-1,2-Cyclohexanedicarboxylic acid ≥98.0%
trans-1,2-Cyclohexanedicarboxylic acid

CAS Number: 2305-32-0
MDL Number: MFCD00001464
Molecular Formula: C8H12O4
Molecular Weight: 172.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 229

Formula: C₈H₁₂O₄
Boiling Pt: 339 °C (1013 hPa)
Melting Pt: 229 °C
MDL Number: MFCD00001464
CAS Number: 2305-32-0
EINECS: 218-975-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=28277d3259051534782262f4119421a7b9e4d280&vid=28277d3259051534782262f4119421a7b9e4d280&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR