Cyclohexanecarboxylic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: Hexahydrobenzoic acid

C0470-500G C0470-25G
TCC0470-500GEA 96.38 CAD
TCC0470-500G TCC0470-025G
Cyclohexanecarboxylic acid ≥98.0% (by titrimetric analysis)
Cyclohexanecarboxylic acid

CAS Number: 98-89-5
MDL Number: MFCD00001461
Molecular Formula: C7H12O2
Molecular Weight: 128.17
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Boiling point (°C): 233
Flash Point (°C): 110
Freezing point (°C): 27

Formula: C₇H₁₂O₂
MW: 128.17 g/mol
Boiling Pt: 233 °C
Melting Pt: 31
Density: 1.03 g/cm³ (20 °C)
Flash Pt: 110
Storage Temperature: Ambient
MDL Number: MFCD00001461
CAS Number: 98-89-5
EINECS: 202-711-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=40df249d3b16ca0d417e270f03c152bf9d2424f7&vid=dd9011e9a0f5bf68fe03fda0b010468889fe4ec0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR