Cyanuric acid ≥98.0%

Supplier: TCI America

Synonyms: 1,3,5-Triazinetrion, 1,3,5-Triazine-2,4,6-triol, 1,3,5-Triazinane-2,4,6-trione, Isocyanuric acid

C0459-500G C0459-25G
TCC0459-500GEA 89.82 CAD
TCC0459-500G TCC0459-025G
Cyanuric acid ≥98.0%
Cyanuric acid

CAS Number: 108-80-5
MDL Number: MFCD00082990
Molecular Formula: C3H3N3O3
Molecular Weight: 129.08
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 360


Other Product Names / Synonyms: Isocyanuric Acid

Formula: C₃H₃N₃O₃
MW: 129.08 g/mol
Melting Pt: 360 °C
Density: 2.5 g/cm³ (20 °C)
MDL Number: MFCD00082990
CAS Number: 108-80-5
EINECS: 203-618-0
Merck Index: 13,02728
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee52dc82eb9ccc79d7b43dab1c6c7718abb8f78f&vid=ee52dc82eb9ccc79d7b43dab1c6c7718abb8f78f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR