cis-1,2-Cyclohexanedicarboxylic acid ≥98.0%
Supplier: TCI America
Synonyms:
cis-Cyclohexane-1,2-dicarboxylic acid
C0458-25G
C0458-500G
TCC0458-025GEA
42.66
CAD
TCC0458-025G
TCC0458-500G
cis-1,2-Cyclohexanedicarboxylic acid ≥98.0%
cis-1,2-Cyclohexanedicarboxylic acid
CAS Number: 610-09-3
MDL Number: MFCD00064949
Molecular Formula: C8H12O4
Molecular Weight: 172.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 198
Flash Point (°C): 250
Formula:
C₈H₁₂O₄ Boiling Pt: 339 °C (1013 hPa) Melting Pt: 198 °C Flash Pt: 250 °C |
MDL Number:
MFCD00064949 CAS Number: 610-09-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...