Isonicotinonitrile ≥98.0%

Supplier: TCI America
Warning

Synonyms: 4-Pyridinecarbonitrile, Pyridine-4-carbonitrile, 4-Cyanopyridine

C0457-25G C0457-500G C0457-100G
TCC0457-025GEA 25.26 CAD
TCC0457-025G TCC0457-500G TCC0457-100G
Isonicotinonitrile ≥98.0%
Isonicotinonitrile

CAS Number: 100-48-1
MDL Number: MFCD00006417
Molecular Formula: C6H4N2
Molecular Weight: 104.11
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 198
Melting point (°C): 80
Flash Point (°C): 92


Product Categories: Synthetic Organic Chemistry, Building Blocks ([2+2+2] Cycloaddition Reaction)
Other Product Names / Synonyms: Isonicotinonitrile

Formula: C₆H₄N₂
Boiling Pt: 198 °C
Melting Pt: 80 °C
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00006417
CAS Number: 100-48-1
EINECS: 202-856-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=76c7e8875629ff891fd8d734771e105e0e3b047c&vid=76c7e8875629ff891fd8d734771e105e0e3b047c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR