Nicotinonitrile ≥98.0%

Supplier: TCI America
Warning

C0456-500G C0456-25G
TCC0456-500GEA 110.6 CAD
TCC0456-500G TCC0456-025G
Nicotinonitrile ≥98.0%
Nicotinonitrile

CAS Number: 100-54-9
MDL Number: MFCD00006372
Molecular Formula: C6H4N2
Molecular Weight: 104.11
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 201
Melting point (°C): 50
Flash Point (°C): 84


Product Categories: Synthetic Organic Chemistry, Building Blocks ([2+2+2] Cycloaddition Reaction)
Other Product Names / Synonyms: Nicotinonitrile

Formula: C₆H₄N₂
Boiling Pt: 201 °C
Melting Pt: 50 °C
Density: 1.159 g/cm³ (25 °C)
Flash Pt: 84 °C
Storage Temperature: Ambient
MDL Number: MFCD00087130
CAS Number: 100-54-9
EINECS: 202-863-0

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR