Vitamin B12 (cyanocobalamin) ≥95.0%
Supplier: TCI America
C0449-1G
C0449-100MG
TCC0449-001GEA
289.76
CAD
TCC0449-001G
TCC0449-100MG
Vitamin B12 (cyanocobalamin) ≥95.0%
Vitamin B12 (cyanocobalamin)
CAS Number: 68-19-9
MDL Number: MFCD00151092
Molecular Formula: C63H88CoN14O14P
Molecular Weight: 1355.39
Purity/Analysis Method: >95.0% (E)
Form: Crystal
Storage Temperature: 0-10°C
Product Categories: Bioscience, Porphyrins, Vitamins
Other Product Names / Synonyms: Cyanocobalamin
Formula:
C₆₃H₈₈CoN₁₄O₁₄P MW: 1355.38 g/mol Melting Pt: 320 Storage Temperature: Refrigerator |
MDL Number:
MFCD00151092 CAS Number: 68-19-9 EINECS: 200-680-0 Merck Index: 12,10152 |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...