4-Cyanobenzoic acid ≥98.0%

Supplier: TCI America
Warning

C0445-5G C0445-25G
TCC0445-005GEA 42.66 CAD
TCC0445-005G TCC0445-025G
4-Cyanobenzoic acid ≥98.0%
4-Cyanobenzoic acid

CAS Number: 619-65-8
MDL Number: MFCD00002528
Molecular Formula: C8H5NO2
Molecular Weight: 147.13
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 224

Formula: C₈H₅NO₂
MW: 147.13 g/mol
Boiling Pt: 313
Melting Pt: 224 °C
Density: 1.382 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002528
CAS Number: 619-65-8
EINECS: 210-606-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d150b7de58823b334fc48360916be15429c5989&vid=7f8a3eed3adf8c5562f28f790017b8164cbd2042&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR