Methyl cyanoacetate ≥99.0%
Supplier: TCI America
Synonyms:
Methyl 2-cyanoacetate
C0442-500ML
C0442-25ML
TCC0442-500MLEA
120.08
CAD
TCC0442-500ML
TCC0442-025ML
Methyl cyanoacetate ≥99.0%
Methyl cyanoacetate
CAS Number: 105-34-0
MDL Number: MFCD00001939
Molecular Formula: C4H5NO2
Molecular Weight: 99.09
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 205
Flash Point (°C): 105
Specific Gravity (20/20): 1.13
Other Product Names / Synonyms: Cyanoacetic Acid Methyl Ester
Formula:
C₄H₅NO₂ MW: 99.09 g/mol Boiling Pt: 207 Melting Pt: -22.5 Density: 1.13 (20 °C) Flash Pt: 105 °C |
MDL Number:
MFCD00001939 CAS Number: 105-34-0 EINECS: 203-288-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...