Ethyl cyanoacetate ≥99.0%
Supplier: TCI America
C0441-25G
C0441-500G
TCC0441-025GEA
42.19
CAD
TCC0441-025G
TCC0441-500G
Ethyl cyanoacetate ≥99.0%
Ethyl cyanoacetate
CAS Number: 105-56-6
MDL Number: MFCD00001940
Molecular Formula: C5H7NO2
Molecular Weight: 113.12
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 206
Melting point (°C): -22
Flash Point (°C): 110
Specific Gravity (20/20): 1.06
Product Categories: Synthetic Organic Chemistry, Cyanation
Other Product Names / Synonyms: Cyanoacetic Acid Ethyl Ester
Formula:
C₅H₇NO₂ Boiling Pt: 206 °C Melting Pt: –22 °C Density: 1.06 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00001940 CAS Number: 105-56-6 EINECS: 203-309-0 Merck Index: 13,03821 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...