Cresol red ≥98.0% (by HPLC)

Supplier: TCI America

C0406-1G C0406-25G
TCC0406-001GEA 48.16 CAD
TCC0406-001G TCC0406-025G
Cresol red ≥98.0% (by HPLC)
Cresol red

CAS Number: 1733-12-6
MDL Number: MFCD00005878
Molecular Formula: C21H18O5S
Molecular Weight: 382.43
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal


Product Categories: Analytical Science, Indicators (pH)
Other Product Names / Synonyms: o-Cresolsulfonphthalein

Formula: C₂₁H₁₈O₅S
Storage Temperature: Ambient
MDL Number: MFCD00005878
CAS Number: 1733-12-6
EINECS: 217-064-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=884b1ac37feea83348b2f6976db7da43903c2b7d&vid=884b1ac37feea83348b2f6976db7da43903c2b7d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR