o-Cresolphthalein

Supplier: TCI America

C0404-25G C0404-100G
TCC0404-025GEA 104.28 CAD
TCC0404-025G TCC0404-100G
o-Cresolphthalein
o-Cresolphthalein

CAS Number: 596-27-0
MDL Number: MFCD00005912
Molecular Formula: C22H18O4
Molecular Weight: 346.38
Form: Crystal
Color: White
Melting point (°C): 225
Lambda max.: 566 nm (pH 9.8 buffer sol.)

Formula: C₂₂H₁₈O₄
Melting Pt: 225 °C
Storage Temperature: Ambient
MDL Number: MFCD00005912
CAS Number: 596-27-0
EINECS: 209-881-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e537496f76c1d124b58a17eff3ab6eb71714ea7c&vid=e537496f76c1d124b58a17eff3ab6eb71714ea7c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR