Creatinine ≥99.0% (by HPLC, titration analysis)

Supplier: TCI America

C0398-25G C0398-500G C0398-100G
TCC0398-025GEA 77.31 CAD
TCC0398-025G TCC0398-500G TCC0398-100G
Creatinine ≥99.0% (by HPLC, titration analysis)
Creatinine

CAS Number: 60-27-5
MDL Number: MFCD00059730
Molecular Formula: C4H7N3O
Molecular Weight: 113.12
Purity/Analysis Method: >99.0% (HPLC,T)
Form: Crystal
Melting point (°C): 255
Flash Point (°C): 290

Formula: C₄H₇N₃O
Melting Pt: 255 °C
Flash Pt: 290 °C
MDL Number: MFCD00059730
CAS Number: 60-27-5
EINECS: 200-466-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea0846f446b932a0771d2a55ababdbb601ee339a&vid=ea0846f446b932a0771d2a55ababdbb601ee339a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR