trans-2-Hydroxycinnamic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: (E)-3-(2-Hydroxyphenyl)acrylic acid, 2-Hydroxycinnamic acid, predominantly trans, trans-2-Hydroxycinnamic acid, trans-o-Coumaric acid, o-Coumaric acid

C0394-5G C0394-25G
TCC0394-005GEA 59.27 CAD
TCC0394-005G TCC0394-025G
trans-2-Hydroxycinnamic acid ≥98.0% (by GC, titration analysis)
trans-2-Hydroxycinnamic acid

CAS Number: 614-60-8
MDL Number: MFCD00004379
Molecular Formula: C9H8O3
Molecular Weight: 164.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 217


Product Categories: Bioscience, Phenylpropanoids, Phenylpropanoids (Phytochemicals)
Other Product Names / Synonyms: trans-o-Cumaric Acid, trans-2-Hydroxycinnamic Acid

Formula: C₉H₈O₃
MW: 164.16 g/mol
Boiling Pt: 330
Melting Pt: 217
MDL Number: MFCD00004379
CAS Number: 614-60-8
EINECS: 210-386-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8040c568a32de4da689ad88da669d961397045a3&vid=86b0604a76d1d2f16626e2fb4fe35aad6d0cb1b2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR