Copper(II) acetylacetonate ≥97.0%

Supplier: TCI America
Warning

Synonyms: Bis(2,4-pentanedionato-O,O')copper (II), Copper (II) bis(2,4-pentanedionato-O,O'), Bis(acetylacetone)copper, Copper (II) 2,4-pentanedionate

C0384-250G C0384-25G
TCC0384-250GEA 158.63 CAD
TCC0384-250G TCC0384-025G
Copper(II) acetylacetonate ≥97.0%
Copper (II) acetylacetonate

CAS Number: 13395-16-9
MDL Number: MFCD00000016
Molecular Formula: C10H14CuO4
Molecular Weight: 261.76
Purity/Analysis Method: >97.0% (T)
Boiling point (°C): 160
Melting point (°C): 288


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Activation Reaction of Unactivated Bonds, Cross-coupling Reaction using Transition Metal Catalysts, Materials Chemistry, Magnetic Metal Complexes (Magnetic Materials)
Other Product Names / Synonyms: Acetylacetone Copper(II) Salt, Copper(II) Acetylacetonate, Cupric Acetylacetonate

Formula: C₁₀H₁₄CuO₄
MW: 261.76 g/mol
Boiling Pt: 160 °C
Melting Pt: 288 °C
MDL Number: MFCD00000016
CAS Number: 13395-16-9
EINECS: 236-477-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ca83eb08990e0adc8c5116a446a83c1fd588f282&vid=ca83eb08990e0adc8c5116a446a83c1fd588f282&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR