ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na, oxidized form), white ≥90.0%, crystals for biochemistry

Supplier: TCI America

Synonyms: NADP monosodium salt, NADP Sodium, Nadide phosphate monosodium salt, Sodium (2R,3R,4R,5R)-2-(6-amino-9H-purin-9-yl)-5-((((((((2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methoxy)oxidophosphoryl)oxy)(hydroxy)phosphoryl)oxy)methyl)-4-hydroxytetrahydrofuran-3-yl hydrogen phosphate, Nicotinamide-adenine dinucleotide phosphate monosodium salt, NADP sodium salt, NADP-Na

C0379-1G C0379-100MG
TCC0379-1GEA 358.2 CAD
TCC0379-1G TCC0379-100MG
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na, oxidized form), white ≥90.0%, crystals for biochemistry
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na, oxidized form)

CAS Number: 1184-16-3
Molecular Formula: C21H29N7O17P3
Molecular Weight: 766.40
Form: Crystals
Color: White


Product Categories: BioscienceNucleosides, Nucleotides, Nucleic Acids, Reagents for Bioanalytical Research, Vitamins

Formula: C₂₁H₂₇N₇NaO₁₇P₃
MW: 765.39 g/mol
Melting Pt: 175 to 178 °C
MDL Number: MFCD00036973
CAS Number: 1184-16-3
EINECS: 214-664-6

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (HPLC)
Melting point (°C) 178
Storage Temperature 0-10°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR