Pyridoxal-5'-phosphate monohydrate ≥98.0%

Supplier: TCI America

Synonyms: Pyridoxal-5-phosphate hydrate, Pyridoxal 5-phosphate monohydrate, Pyridoxal 5'-phosphate monohydrate, (4-Formyl-5-hydroxy-6-methylpyridin-3-yl)methyl dihydrogen phosphate hydrate

C0377-25G C0377-1G C0377-5G
TCC0377-025GEA 493.29 CAD
TCC0377-025G TCC0377-001G TCC0377-5G
Pyridoxal-5'-phosphate monohydrate ≥98.0%
Pyridoxal 5'-phosphate monohydrate

CAS Number: 41468-25-1
MDL Number: MFCD00006333
Molecular Formula: C8H10NO6P
Molecular Weight: 247.14
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 143


Product Categories: Bioscience, Vitamin Derivatives
Other Product Names / Synonyms: Codecarboxylase, PLP

Formula: C₈H₁₀NO₆P·1H₂O
Melting Pt: 143 °C
Storage Temperature: Freezer
MDL Number: MFCD00149414
CAS Number: 41468-25-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c088dee220184de3247cce9b00aacaf1f95897ed&vid=c088dee220184de3247cce9b00aacaf1f95897ed&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR