(±)-β-Citronellol ≥92.0%

Supplier: TCI America

C0370-500ML C0370-25ML C0370-100ML
TCC0370-500MLEA 150.1 CAD
TCC0370-500ML TCC0370-025ML TCC0370-100ML
(±)-β-Citronellol ≥92.0%
(±)-β-Citronellol

CAS Number: 106-22-9
MDL Number: MFCD00002935
Molecular Formula: C10H20O
Molecular Weight: 156.27
Purity/Analysis Method: >92.0% (GC)
Form: Clear Liquid
Boiling point (°C): 220
Flash Point (°C): 102
Specific Gravity (20/20): 0.86


Product Categories: Bioscience, Reagents for Inflammation & Immunity Disorder Models, Acyclic Monoterpenes
Other Product Names / Synonyms: 3,7-Dimethyl-6-octen-1-ol

Formula: C₁₀H₂₀O
Boiling Pt: 220 °C
Density: 0.86 (20 °C)
Flash Pt: 102 °C
MDL Number: MFCD00002935
CAS Number: 106-22-9
EINECS: 203-375-0
UN: 3082
ADR: 9,III
Merck Index: 13,02354
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b72c335a4fb2092662eef87f98cc5d2cf9afd939&vid=b72c335a4fb2092662eef87f98cc5d2cf9afd939&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 92.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR