Tributyl citrate ≥98.0%
Supplier: TCI America
C0366-25ML
C0366-500ML
TCC0366-025MLEA
34.76
CAD
TCC0366-025ML
TCC0366-500ML
Tributyl citrate ≥98.0%
Tributyl citrate
CAS Number: 77-94-1
MDL Number: MFCD00027217
Molecular Formula: C18H32O7
Molecular Weight: 360.45
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 233
Melting point (°C): -20
Flash Point (°C): 206
Specific Gravity (20/20): 1.04
Product Categories: Materials Chemistry, Resin Additives
Other Product Names / Synonyms: Butyl Citrate, Citric Acid Tributyl Ester
Formula:
C₁₈H₃₂O₇ Boiling Pt: 233 °C Melting Pt: –20 °C Density: 1.04 (20 °C) Flash Pt: 206 °C Storage Temperature: Ambient |
MDL Number:
MFCD00027217 CAS Number: 77-94-1 EINECS: 201-071-2 Merck Index: 13,01563 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...