Citraconic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

C0363-25G C0363-500G
TCC0363-025GEA 71.1 CAD
TCC0363-025G TCC0363-500G
Citraconic acid ≥98.0% (by GC, titration analysis)
Citraconic acid

CAS Number: 498-23-7
MDL Number: MFCD00078086
Molecular Formula: C5H6O4
Molecular Weight: 130.10
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 87


Other Product Names / Synonyms: Methylmaleic Acid

Formula: C₅H₆O₄
Boiling Pt: 336 °C (1013 hPa)
Melting Pt: 87 °C
Density: 1.617 g/cm³ (20 °C)
MDL Number: MFCD00078086
CAS Number: 498-23-7
EINECS: 207-858-7
Merck Index: 13,02345
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=25628ec3938b8c62b91ed35ba8665d57d4be29d7&vid=25628ec3938b8c62b91ed35ba8665d57d4be29d7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR