Cinnamyl alcohol ≥97.0%

Supplier: TCI America

C0362-25G C0362-500G
TCC0362-025GEA 35.36 CAD
TCC0362-025G TCC0362-500G
Cinnamyl alcohol ≥97.0%
Cinnamyl alcohol

CAS Number: 104-54-1
MDL Number: MFCD00002921
Molecular Formula: C9H10O
Molecular Weight: 134.18
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 250
Flash Point (°C): 126
Freezing point (°C): 31

Formula: C₉H₁₀O
Boiling Pt: 250 °C
Melting Pt: 31…34 °C
Density: 1.04 g/cm³ (25 °C)
Flash Pt: 126 °C
Storage Temperature: Ambient
MDL Number: MFCD00002921
CAS Number: 104-54-1
EINECS: 203-212-3
Merck Index: 13,02325
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f15329783a0eaf0f84b1449ef2f576197b96de4&vid=3f15329783a0eaf0f84b1449ef2f576197b96de4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR