Methyl cinnamate ≥99.0%

Supplier: TCI America

Synonyms: Methyl 3-phenylpropenoate, Methyl 3-phenyl-2-propenoate

C0360-25G C0360-500G
TCC0360-025GEA 30.02 CAD
TCC0360-025G TCC0360-500G
Methyl cinnamate ≥99.0%
Methyl cinnamate

CAS Number: 103-26-4
MDL Number: MFCD00008458
Molecular Formula: C10H10O2
Molecular Weight: 162.19
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 262
Flash Point (°C): 100
Freezing point (°C): 34


Product Categories: Bioscience, Phenylpropanoids
Other Product Names / Synonyms: Cinnamic Acid Methyl Ester, Methyl trans-3-Phenylacrylate, trans-3-Phenylacrylic Acid Methyl Ester

Formula: C₁₀H₁₀O₂
MW: 162.19 g/mol
Boiling Pt: 262 °C
Flash Pt: 100 °C
MDL Number: MFCD00008458
CAS Number: 103-26-4
EINECS: 203-093-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7eab9a1dbfe5ad006b0ae9b4a26917c41da94b1a&vid=7eab9a1dbfe5ad006b0ae9b4a26917c41da94b1a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR