Ethyl cinnamate ≥99.0%

Supplier: TCI America
Warning

C0359-25G C0359-500G C0359-100G
TCC0359-025GEA 72.68 CAD
TCC0359-025G TCC0359-500G TCC0359-100G
Ethyl cinnamate ≥99.0%
Ethyl cinnamate

CAS Number: 103-36-6
MDL Number: MFCD00009189
Molecular Formula: C11H12O2
Molecular Weight: 176.22
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 271
Melting point (°C): 10
Flash Point (°C): 137
Specific Gravity (20/20): 1.05


Product Categories: Bioscience, Phenylpropanoids
Other Product Names / Synonyms: Cinnamic Acid Ethyl Ester, Ethyl trans-3-Phenylacrylate, trans-3-Phenylacrylic Acid Ethyl Ester

Formula: C₁₁H₁₂O₂
Boiling Pt: 271 °C
Melting Pt: 10 °C
Density: 1.05 (20 °C)
Flash Pt: 137 °C
MDL Number: MFCD00009189
CAS Number: 103-36-6
EINECS: 203-104-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=be1411b4fc0cb42d07251bf6929282b180ad5a84&vid=be1411b4fc0cb42d07251bf6929282b180ad5a84&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR