Benzyl cinnamate ≥98.0%

Supplier: TCI America

Synonyms: Cinnamic acid benzyl ester, trans-Cinnamic acid benzyl ester, 3-Phenyl-2-propenoic acid benzyl ester

C0358-25G C0358-500G
TCC0358-025GEA 30.02 CAD
TCC0358-025G TCC0358-500G
Benzyl cinnamate ≥98.0%
Benzyl cinnamate

CAS Number: 103-41-3
MDL Number: MFCD00004789
Molecular Formula: C16H14O2
Molecular Weight: 238.29
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 230
Melting point (°C): 36
Flash Point (°C): 100


Product Categories: Bioscience, Phenylpropanoids
Other Product Names / Synonyms: Cinnamic Acid Benzyl Ester

Formula: C₁₆H₁₄O₂
MW: 238.29 g/mol
Boiling Pt: 230 °C
Melting Pt: 36 °C
Flash Pt: 100 °C
MDL Number: MFCD00004789
CAS Number: 103-41-3
EINECS: 203-109-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0709ab95624e1b6b350450440fd7764757708daa&vid=0709ab95624e1b6b350450440fd7764757708daa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR