4-Chloro-o-xylene ≥98.0%

Supplier: TCI America

Synonyms: 4-Chloro-1,2-dimethylbenzene, 1-Chloro-3,4-dimethylbenzene

C0356-100G C0356-25G
TCC0356-100GEA 540.36 CAD
TCC0356-100G TCC0356-25G
4-Chloro-o-xylene ≥98.0%
4-Chloro-o-xylene

CAS Number: 615-60-1
MDL Number: MFCD00000596
Molecular Formula: C8H9Cl
Molecular Weight: 140.61
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 192
Flash Point (°C): 66
Specific Gravity (20/20): 1.07


Other Product Names / Synonyms: 3,4-Dimethylchlorobenzene

Formula: C₈H₉Cl
Boiling Pt: 192 °C
Melting Pt: –6 °C
Density: 1.07 (20 °C)
Flash Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD00000596
CAS Number: 615-60-1
EINECS: 210-438-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=43f443f316c62730d761907a0bc808191227fe3c&vid=facd11faa6fede6df5ff5b1e7794f29799d4e5e0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR