trans-Cinnamic acid ≥98.0%

Supplier: TCI America
Warning

Synonyms: Cinnamic acid (only trans), (2E)-2-Phenyl-2-propenoic acid, trans-Cinnamic acid

C0353-25G C0353-500G C0353-100G
TCC0353-025GEA 50.99 CAD
TCC0353-025G TCC0353-500G TCC0353-100G
trans-Cinnamic acid ≥98.0%
trans-Cinnamic acid

CAS Number: 140-10-3
MDL Number: MFCD00004369
Molecular Formula: C9H8O2
Molecular Weight: 148.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 300
Melting point (°C): 135
Lambda max.: 273 nm (MeOH)


Product Categories: Synthetic Organic Chemistry, Carboxylic Acids (Multi-component Reaction), Bioscience, Crude Medicine Ingredients for Research and Experimental Use, Phenylpropanoids, Phenylpropanoids (Phytochemicals), Analytical Science, Standards for Mass Spectrometry &
Other Product Names / Synonyms: trans-3-Phenylacrylic Acid

Formula: C₉H₈O₂
MW: 148.16 g/mol
Boiling Pt: 300
Melting Pt: 135 °C
Density: 1.25 g/cm³ (4 °C)
MDL Number: MFCD00004369
CAS Number: 140-10-3
EINECS: 205-398-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=99fc10c5f81fc4aa86576230911ddc9362fc1bba&vid=99fc10c5f81fc4aa86576230911ddc9362fc1bba&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR