trans-Cinnamaldehyde ≥98.0%
Supplier: TCI America
CAS Number: 14371-10-9
MDL Number: MFCD00007000
Molecular Formula: C9H8O
Molecular Weight: 132.16
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 252
Melting point (°C): -8
Flash Point (°C): 129
Specific Gravity (20/20): 1.05
Product Categories: Synthetic Organic Chemistry, Aldehydes (Multi-component Reaction), Bioscience, Fungicide Ingredients, Crude Medicine Ingredients for Research and Experimental Use, Phenylpropanoids, Phenylpropanoids (Phytochemicals)
Other Product Names / Synonyms: trans-3-Phenylacrolein, trans-3-Phenylacrylaldehyde, trans-3-Phenyl-2-propenal
Formula:
C₉H₈O Boiling Pt: 252 °C Melting Pt: –8 °C Density: 1.05 (20 °C) Flash Pt: 129 °C |
MDL Number:
MFCD00007000 CAS Number: 14371-10-9 Merck Index: 12,02319 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...