2-Chlorobenzothiazole ≥98.0%
Supplier: TCI America
C0332-25G
C0332-250G
TCC0332-025GEA
104.88
CAD
TCC0332-025G
TCC0332-250G
2-Chlorobenzothiazole ≥98.0%
2-Chlorobenzothiazole
CAS Number: 615-20-3
MDL Number: MFCD00005776
Molecular Formula: C7H4ClNS
Molecular Weight: 169.63
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 248
Flash Point (°C): 110
Freezing point (°C): 21
Other Product Names / Synonyms: 2-Benzothiazolyl Chloride
Formula:
C₇H₄ClNS Boiling Pt: 248 °C Melting Pt: 21…24 °C Density: 1.37 g/cm³ (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00005776 CAS Number: 615-20-3 EINECS: 210-415-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...