4-Chlororesorcinol ≥98.0%

Supplier: TCI America
Warning

C0285-500G C0285-25G
TCC0285-500GEA 712.58 CAD
TCC0285-500G TCC0285-025G
4-Chlororesorcinol ≥98.0%
4-Chlororesorcinol

CAS Number: 95-88-5
MDL Number: MFCD00002273
Molecular Formula: C6H5ClO2
Molecular Weight: 144.55
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 256
Melting point (°C): 107


Other Product Names / Synonyms: 1-Chloro-2,4-dihydroxybenzene

Formula: C₆H₅ClO₂
Boiling Pt: 256 °C
Melting Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00002273
CAS Number: 95-88-5
EINECS: 202-462-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9f8eebb894df191fadab0d085ed47658a22c60b7&vid=9f8eebb894df191fadab0d085ed47658a22c60b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR