6-Chloroquinoline ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

C0283-5G C0283-1G
TCC0283-005GEA 251.22 CAD
TCC0283-005G TCC0283-1G
6-Chloroquinoline ≥98.0% (by GC, titration analysis)
6-Chloroquinoline

CAS Number: 612-57-7
MDL Number: MFCD00024024
Molecular Formula: C9H6ClN
Molecular Weight: 163.60
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 264
Melting point (°C): 41
Flash Point (°C): 110

Formula: C₉H₆ClN
MW: 163.61 g/mol
Boiling Pt: 264 °C
Melting Pt: 41 °C
Density: 1.2146 g/cm³ (56 °C)
Flash Pt: 110 °C
MDL Number: MFCD00024024
CAS Number: 612-57-7
EINECS: 210-314-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0f6234f00615c9a15743ddddcfb9577aca9059d&vid=cb8c5c636a9d57a818890544bb02c48ff6075e13&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR