2-Chloroquinoline ≥98.0%
Supplier: TCI America
|
Warning
|
C0282-5G
C0282-25G
TCC0282-005GEA
105.86
CAD
TCC0282-005G
TCC0282-025G
2-Chloroquinoline ≥98.0%
2-Chloroquinoline
CAS Number: 612-62-4
MDL Number: MFCD00006741
Molecular Formula: C9H6ClN
Molecular Weight: 163.60
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 267
Melting point (°C): 36
Flash Point (°C): 110
Storage Temperature: 0-10°C
Formula:
C₉H₆ClN MW: 163.61 g/mol Boiling Pt: 267 °C Melting Pt: 36 °C Density: 1.2464 g/cm³ (25 °C) Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006741 CAS Number: 612-62-4 EINECS: 210-317-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...