3-Chloropyridine ≥95.0%

Supplier: TCI America

C0280-25G C0280-500G C0280-100G
TCC0280-025GEA 61.78 CAD
TCC0280-025G TCC0280-500G TCC0280-100G
3-Chloropyridine ≥95.0%
3-Chloropyridine

CAS Number: 626-60-8
MDL Number: MFCD00006375
Molecular Formula: C5H4ClN
Molecular Weight: 113.54
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 148
Flash Point (°C): 66
Specific Gravity (20/20): 1.21

Formula: C₅H₄ClN
MW: 113.55 g/mol
Boiling Pt: 148 °C
Melting Pt: –0.5 °C
Density: 1.21 (20 °C)
Flash Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD00006375
CAS Number: 626-60-8
EINECS: 210-955-7
UN: 1100
ADR: 3 I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ffdb6c54d1330d274d565c822dea59ac6e5c2795&vid=ffdb6c54d1330d274d565c822dea59ac6e5c2795&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR